Learn More
TARGETMOL CHEMICALS INC RS-1 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T697225MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. RS-1 is a RAD51 activator. RS-1 also increases CRISPR/Cas9-mediated knock-in efficiencies. Purity 99.72%
Catalog No. 50-288-0096