Learn More
Medchemexpress LLC HY-19793 5mg Medchemexpress, RS-1 CAS:312756-74-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY197935MG
Encompass_Preferred
Medchemexpress, HY-19793 5mg RS-1 CAS:312756-74-4 RS-1 is a RAD51 activator, and also increases CRISPR/Cas9-mediated knock-in efficiencies. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2185