Learn More
Abcam RS 102221 hydrochloride, 5-HT2C receptor antagonist, 10MG
Supplier:  Abcam AB12098210MG
Encompass_Preferred
MW 649.08 g/mol, Purity >98%. Selective, high affinity 5-HT₂C receptor antagonist (pKi = 8.4). Displays approximately 100-fold selectivity over the 5-HT₂A and 5-HT₂B subtypes and > 100-fold selectivity over other 5-HT receptors, α- and β-adrenergic and muscarinic ACh receptors.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7064