Learn More
Selleck Chemical LLC RS-102895 Hydrochloride
SDP
Supplier:  Selleck Chemical LLC S753825MG
Encompass_Preferred
RS-102895 hydrochloride (HCl) is a potent antagonist of Chemokine (C-C motif) receptor 2 (CCR2) with IC50 of 360 nM and shows no effect on CCR1 RS-102895 hydrochloride also inhibits human 1a and 1d receptors rat brain cortex 5-HT1a receptor in cells with IC50s of 130 nM 320 nM 470 nM respectively
Catalog No. 50-000-04746