Learn More
TARGETMOL CHEMICALS INC RS 17053 HYDROCHLORIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T167985MG
Encompass_Preferred
Also available in 2 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RS 17053 hydrochloride is a potent and selective antagonist of (alpha)1A adrenoceptor. RS 17053 hydrochloride exhibits a pKi value of 9.1 in native cell membrane and a pA2 value of 9.8 in functional assays. purity: 99%
Catalog No. 50-258-1151