Learn More
TARGETMOL CHEMICALS INC RS-4-CARBOXYPHENYLGLYCIN 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T232625MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. (RS)-4-Carboxyphenylglycine is a racemic mixture. (S)-4-Carboxyphenylglycine is a selective mGlu1(alpha) receptor antagonist. purity: 99%
Catalog No. 50-257-7158