Learn More
Medchemexpress LLC RS 67333 hydrochloride | 168986-60-5 | 99.7% | 389.36 g/mol | C19H30Cl2N2O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10134150MG
Encompass_Preferred
RS 67333 hydrochloride is the hydrochloride salt of a selective partial agonist of the 5-HT4 (serotonin) receptor. Supplied as a solid research reagent, it is used in neuroscience studies of 5-HT4 receptor pharmacology, neuroprotection, and related pathways. For research use only.
  • Selective partial agonist of 5-HT4 receptor.
  • High purity (99.7%).
  • Supplied as a solid suitable for in vitro and in vivo studies.
  • Stable when stored sealed at 4°C, away from moisture.
  • Available in small laboratory quantities for research applications.
Catalog No. 50-002-11958