Learn More
Abcam (R,S)-CPP, NMDA antagonist, 50MG
Supplier:  Abcam AB12016050MG
Encompass_Preferred
MW 252.2 Da. Potent, competitive NMDA antagonist. See separate isomer (ab120159). Also available in simple stock solutions (ab146720) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7834