Learn More
Medchemexpress LLC (RS)-CPPG | 183364-82-1 | 99.0% | 271.21 g/mol | C11H14NO5P | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1013335MG
Encompass_Preferred
CPPG ((RS)-CPPG) is a research-grade antagonist of group II and III metabotropic glutamate receptors used in pharmacology and neuroscience studies. It is a white solid with high purity, molecular formula C11H14NO5P, molecular weight 271.21 g/mol, and CAS number 183364-82-1. Store powdered material at -20°C for long-term stability.
  • High purity suitable for research applications.
  • Selective antagonist of group II and III mGlu receptors.
  • Powder form facilitates storage and formulation.
  • Stable under recommended storage conditions.
  • Suitable quantities for screening and assay work.
Catalog No. 50-002-11413