Learn More
Abcam (R,S)-Lenalidomide, Thalidomide analog, TNF alpha inhibitor, 100MG
Supplier:  Abcam AB142129100MG
Encompass_Preferred
MW 259.26 Da, Purity >98%. Thalidomide (ab120032) analog and TNF-α inhibitor. Potent antitumor and anti-inflammatory effects. Modulates and potentiates host immune responses against multiple myeloma. Active in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6321