Learn More
Medchemexpress LLC (RS)-(Tetrazol-5-yl)glycine | 138199-51-6 | MFCD00233633 | 96.5% | 143.10 g/mol | C3H5N5O2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1008391MG
Encompass_Preferred
(RS)-(Tetrazol-5-yl)glycine is a research-grade NMDA receptor agonist used to activate NMDA-type glutamate receptors in neuropharmacology and electrophysiology studies. Supplied as a solid with documented purity and stability data, it is intended for laboratory research applications and is not for human or clinical use.
  • Selective NMDA receptor agonist for receptor pharmacology and electrophysiology.
  • High purity suitable for biochemical and cellular assays.
  • Available in small milligram quantities for dose-response experiments.
  • Solid form with recommended low-temperature storage to preserve stability.
  • Accompanied by data sheet and certificate of analysis for quality verification.
Catalog No. 50-002-12343