Learn More
TARGETMOL CHEMICALS INC RS102895 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TQ028325MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RS102895 is a potent CCR2 antagonist (IC50 360 nM) and shows no effect on CCR1. purity: 99%
Catalog No. 50-257-8804