Learn More
MilliporeSigma Reference Standard Endotoxin,
Supplier:  MilliporeSigma 1441610001
Encompass
The Reference Standard Endotoxin (RSE) is the endotoxin standard recommended by the European Pharmacopeia and supplied in a lyophilized form. Its assigned content is 10 000 International Units per vial.
Catalog No. NC1581381