Learn More
NABERTHERM INC These tube furnaces can be used for horizontal operation. The split-type design makes it easy to change the working tube. It allows for a comfortable exchange of various working tubes.
Supplier:  NABERTHERM INC RS252L3AN240
Encompass_Preferred
These tube furnaces can be used for horizontal operation. The split-type design makes it easy to change the working tube. It allows for a comfortable exchange of various working tubes (e.g. working tubes made of different materials).Using a wide range of accessories, these professional tube furnaces can be optimally tailored for your process. By adding different gas supply packages, you can work in a protective gas atmosphere, with gases or in a vacuum. In addition to the convenient standard controllers, modern PLC controls can also be used to control the process.
Catalog No. NC2332310
Add to Cart
Request a Quote