Learn More
Medchemexpress LLC RSK4-IN-1 trifluoroacetate | 00-00-0 | 98.6% | 390.38 g/mol | C19H20F2N4O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY132891A5MG
Encompass_Preferred
RSK4-IN-1 TFA is the trifluoroacetate salt of a potent small-molecule inhibitor of ribosomal S6 kinase 4 (RSK4). It inhibits RSK4 with an IC50 of 9.5 nM and has reported antitumor activity. Supplied as a research-grade compound with high purity, it is intended for biochemical and cellular studies of RSK4 signaling.
  • Potent RSK4 inhibition (IC50 9.5 nM).
  • Reported antitumor activity in preclinical studies.
  • High purity (98.6%).
  • Molecular formula C19H20F2N4O3; molecular weight 390.38 g/mol.
  • Supplied as a trifluoroacetate salt suitable for biochemical and cellular assays.
  • Available in milligram pack sizes (1 mg-100 mg) for research use.
Catalog No. 50-002-62386