Learn More
Drg International Inc DRG RSV IgM ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3508R
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgM class antibodies to Respiratory Syncytial Virus (RSV) in serum. This product is for Research Use Only. The DRG RSV IgM ELISA is a solid phase enzyme-linked immunosorbent assay (ELISA). Samples are diluted with Sample Diluent and additionally incubated with IgG-RF-Sorbent, containing hyper-immune anti-human IgG class antibody to eliminate competitive inhibition from specific IgG and to remove rheumatoid factors. This pretreatment avoids false results. Microtiter wells as a solid phase are coated with RSV antigen. Pretreated specimens and ready for use controls are pipetted into these wells.
Catalog No. 50-194-7585