Learn More
Sino Biological RSV RSV-F ELISA Pair Set 15PP (Sino Biological)
Supplier:  Sino Biological SEK1104915
Encompass_Preferred
Each vial contains 203 ng of recombinant Human respiratory syncytial virus (RSV) Fusion Glycoprotein / RSV-F. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial in Dilution Buffer, and a high standard of 8000 pg/mL is recommended.
Catalog No. 50-237-3058