Learn More
BIOGEMS INTERNATIONAL INC Rat IgG2a Isotype Control FITC Size 100 g
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 4181350100
Encompass
The 2A3 antibody was obtained by hybridization of MouseNS-1 myeloma cells with spleen cells harvested from BALB/c mice immunized with phytohemagglutinin-activated human T cells. The 2A3 antibody is a CD25 clone and used as an Isotype Control for rat IgG2a antibodies.
Catalog No. 50-153-0087