Learn More
Biocare Medical, LLC RT-ON-MOUSE AP-POLYMER 25ML
SDP
Supplier:  Biocare Medical, LLC RT518H
Encompass_Preferred
The Rat-on-Mouse AP-Polymer is for use with rat primary antibodies on mouse tissues. This detection system is mouse adsorbed for minimum cross-reactivity to endogenous mouse IgG. It is labeled with alkaline phosphatase (AP).
Catalog No. 50-832-88