Learn More
Biocare Medical, LLC RT-ON-MOUSE HRP-POLYMER 100ML
SDP
Supplier:  Biocare Medical, LLC RT517L
Encompass_Preferred
The Rat-on-Mouse HRP-Polymer is for use with rat primary antibodies on mouse tissues. This detection system is mouse adsorbed for minimum cross-reactivity to endogenous mouse IgG. It is labeled with horseradish peroxidase (HRP).
Catalog No. 50-832-85