Learn More
Biocare Medical, LLC Rat-on-Mouse HRP (horseradish peroxidase)-Polymer, 25ml
SDP
Supplier:  Biocare Medical, LLC RT517H
Encompass_Preferred
The Rat-on-Mouse HRP-Polymer is for use with rat primary antibodies on mouse tissues. This detection system is mouse adsorbed for minimum cross-reactivity to endogenous mouse IgG. It is labeled with horseradish peroxidase (HRP).
Catalog No. 50-832-84