Learn More
PREDICT HEALTH HIV viral load (Quantitative) RT-PCR kit is a quantitative RT PCR based diagnostic kit for detection of HIV RNA in human plasma/serum or blood samples. 100 reactions
Supplier:  PREDICT HEALTH DVZ 30081
Encompass_Preferred
HIV viral load (Quantitative) RT-PCR kit is an in vitro nucleic acid amplification test for the detection & quantitation of the HIV RNA in human plasma/serum or blood samples. This test kit utilizes the real time RT-PCR after the extraction of pure nucleic acid from the plasma/serum samples. The viral load is measured using a range of four quantification calibrators provided in the kit.
Catalog No. NC2891004