Learn More
Aviva Systems Biology Corp RTP1 Antibody - C-terminal region (ARP65493_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65493_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RTP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human RTP1. Alternate names: MGC35450ApplicationWB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-288-507