Learn More
Vector Laboratories R.T.U. Horseradish Peroxidase Streptavidin (Ready-To-Use)
SDP
Supplier:  Vector Laboratories SA5704100
Encompass
Avidin and streptavidin reagents are powerful tools for the detection or purification of biotinylated proteins, nucleic acids, and other macromolecules. This Horseradish Peroxidase Streptavidin (HRP Streptavidin) is produced using a proprietary coupling procedure that preserves the high specific activity of the peroxidase. Using it with biotinylated primary or secondary intermediates can help localize antigens in histological sections, cytospin preparations, or smears. This Horseradish Peroxidase Streptavidin is supplied as a ready-to-use (R.T.U.) stabilized solution (100 ml, 1 g/ml) in a bottle fitted with a drop dispenser top. It is also available as a concentrate (1mg/ml) for use in solid-phase assays such as ELISAs and blotting applications.
Catalog No. NC9719944