Learn More
Abcam RU 24969 hemisuccinate, 5-HT1A/1B agonist, 50MG
Supplier:  Abcam AB14629950MG
Encompass_Preferred
MW 574.7 Da, Purity >99%. Potent 5-HT1A/1B agonist (ED50 = 236 nM). Locomotor-activating effects. Centrally active. Active *in vitro* and *in vivo.*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4746