Learn More
Medchemexpress LLC Ru-301 | 1110873-99-8 | 99.8% | 480.46 g/mol | C21H19F3N4O4S | 1 ML

Supplier: Medchemexpress LLC HY11903910MM 1ML
RU-301 is a small-molecule pan-TAM (Tyro3/Axl/Mer) receptor inhibitor supplied as a 10 mM solution in DMSO for research use in cell signaling and oncology. It is provided at high reported purity with recommended storage conditions for both powder and solution forms.
- Pan-TAM receptor inhibitor that blocks Gas6-induced TAM activation.
- Supplied as a 10 mM solution in DMSO for convenient handling.
- Reported high purity suitable for biochemical and cell-based assays.
- Molecular weight 480.46 g/mol with formula C21H19F3N4O4S.
- Storage recommendations provided for powder and in-solvent conditions to maintain stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.