Learn More
Medchemexpress LLC Ru-302 | 1182129-77-6 | 98.7% | 475.53 g/mol | C24H24F3N3O2S | 25MG
SDP
Supplier:  Medchemexpress LLC HY12406625MG
Encompass_Preferred
RU-302 is a small-molecule pan-TAM receptor inhibitor that disrupts the interaction between the TAM Ig1 ectodomain and the Gas6 LG domain, inhibiting Gas6-inducible Axl activation in cell assays and showing antitumor activity in lung cancer models. Supplied as a high-purity solid with solution formats available for research use only.
  • Pan-TAM receptor inhibitor targeting Tyro3, Axl, Mer
  • Blocks the TAM Ig1-Gas6 LG interaction
  • Shows cell-based activity at low micromolar IC50
  • High supplied purity (~98.7%)
  • Available as solid and DMSO solution in multiple sizes
  • For research use only; not for human consumption
Catalog No. 50-002-38324