Learn More
Medchemexpress LLC RU-302 | 1182129-77-6 | 98.7% | 475.53 | C24H24F3N3O2S | 10MG
SDP
Supplier:  Medchemexpress LLC HY12406610MG
Encompass_Preferred
RU-302 is a pan-TAM receptor inhibitor that blocks the interface between the TAM Ig1 ectodomain and the Gas6 Lg domain, inhibiting Gas6-inducible Axl receptor activation in cell assays and showing suppression of lung cancer tumor growth. Supplied as a research-grade small molecule, it is intended for laboratory use only.
Catalog No. 50-002-36750