Learn More
Apexbio Technology LLC RU.521 2262452-06-0 5mg
SDP
Supplier:  Apexbio Technology LLC B89065
Encompass_Preferred
RU 521 (CAS 2262452-06-0) is a selective inhibitor of cyclic GMP-AMP synthase (cGAS) an enzyme responsible for synthesizing cGAMP upon recognition of cytosolic double-stranded DNA By interfering with cGAS-mediated cGAMP production RU 521 suppresses activation of the type I interferon signaling pathway Experimental studies demonstrate that RU 521 reduces cGAS-dependent signal transduction and interferon responses in reporter assays containing double-stranded DNA In macrophages derived from Aicardi-Gouti res syndrome mouse models RU 521 lowers basal interferon expression This compound is a valuable tool for investigating cGAS-dependent innate immune pathways
Catalog No. 50-000-89335