Learn More
Cayman Chemical RU-SKI 201hydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 211015 MG
Encompass_Preferred
An Hhat inhibitor (IC50 = 0.2 μM for recombinant enzyme); inhibits palmitoylation of Shh in Shh-overexpressing HEK293 cells with a tagging IC50 (TC50) value of 0.87 μM; inhibits Hh signaling in H520, PANC-1, and MCF-7 cancer cells co-cultured with Shh-Light2 cells (IC50s = 4.8, 7.8, and 8.5 μM, respectively)
Catalog No. 50-298-3488