Learn More
Medchemexpress LLC Ru-ski 43 hydrochloride | 1782573-67-4 | 99.6% | 423.01 g/mol | C22H31ClN2O2S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY18366A10MG
Encompass_Preferred
RU-SKI 43 hydrochloride is the hydrochloride salt of a small-molecule inhibitor of hedgehog acyltransferase (Hhat) used in research to probe hedgehog signaling and evaluate anticancer effects in cellular models.
  • Potent and selective Hhat inhibitor (IC50 850 nM).
  • Useful for hedgehog pathway and oncology research.
  • Reduces hedgehog signaling and Shh palmitoylation in cellular assays.
  • High purity suitable for biochemical and cell-based studies (~99.6%).
  • Characterized by formula C22H31ClN2O2S and molecular weight 423.01 g/mol.
Catalog No. 50-002-95266