Learn More
Electron Microscopy Sciences EMS High Precision Rubber Grip Cutting Tweezer Style 14AGW, Angled Blade, 115 mm OAL
SDP
Supplier:  Electron Microscopy Sciences 7832508
Encompass_Preferred
Soft ESD safe ergonomic cushion grips (resistivity 10^8-10^9 Ohm) provide enhanced operator comfort and reduced finger stress when handling ESD sensitive components or small static items. Cutting tweezers have fine tips, max visibility, predominantly angled blades.
Blade: Angled
OAL: 115 mm
Catalog No. 50-193-1004