Learn More
Cole-Parmer Cole-Parmer Valved Pipette Aspirator/Bulb/Filler; 50 mL capacity
Supplier:  Cole-Parmer 2480510
Encompass_Preferred
Cole-Parmer Valved Pipette Filler fits standard pipettes and has a 50 mL capacity. One-hand control of three air valves lets you evacuate bulb, fill pipette, and dispense liquid easily.
Catalog No. NC9766310