Learn More
Enterprise Technology Solutions Rubbermaid Commercial Products Air Freshener Refill,f/T-Cell Dispenser,Mango Blossom
SDP
Supplier:  Enterprise Technology Solutions RCP402369
Encompass_Preferred
T-Cell air freshener refill is part of an odor-control system that dispenses a consistent level of fragrance and Microtrans odor neutralizer. Through a Rubbermaid T-Cell dispenser, the fresh fragrance and odor neutralizer are distributed throughout the room. Mango blossom is a natural and exotic fruity fragrance characterized by ripe, juicy mango slices alongside fresh citrus and soft floral accents. Refill lasts approximately 60 days on the standard setting for a washroom with three to four stalls/fixtures. It can last approximately 90 days in a small washroom with one to two stalls/fixtures. Refill contains no propellants or added VOCs.
Catalog No. 50-007-90793