Learn More
Enterprise Technology Solutions Rubbermaid Commercial Products MB 9000 Air Spray Refill, 4/CT, Ocean Breeze
SDP
Supplier:  Enterprise Technology Solutions RCP4012471
Encompass_Preferred
Ocean Breeze Refill delivers long-lasting, effective odor control in Rubbermaid Commercial Microburst 9000 Dispensers. While traditional air fresheners only camouflage malodors, this refill is engineered to do the complete job with Microtrans, a true odor neutralizer. It is paired with high-quality fragrances to effectively eliminate unavoidable odors in commercial washrooms. Ocean Breeze features refreshing citrus and herbal notes that are topped with florals on a musk base. Refill actuator distributes the industry's finest mist for longer-lasting fragrance with 9,000 sprays per refill. Refill contains 63 percent less VOCs per spray than a standard aerosol.
Catalog No. 50-007-90952