Learn More
Enterprise Technology Solutions Rubbermaid Commercial Products Over-The-Spill Station Kit,w/Dispenser&Pads&Fasteners,6/CT,Yellow
SDP
Supplier:  Enterprise Technology Solutions RCP425100YEL
Encompass_Preferred
Slip-resistant pads quickly cover spills and immediately warn pedestrians of a walking hazard in both English and Spanish. The pad also displays the universal caution symbol on a bright yellow background. It absorbs up to 16 oz. of spilled liquids, including water, oils and most solvents. Pads are made of polypropylene for added strength and absorbency. Over-the-Spill Station Kit includes pad dispenser, 22 large pads and fasteners. Dispensing station mounts on flat surfaces. Floor sign stands are sold separately.
Catalog No. 50-008-29543