Learn More
Enterprise Technology Solutions Rubbermaid Commercial Products Toilet Bowl Brush, Bristles, 17" Long, Brown/Yellow
SDP
Supplier:  Enterprise Technology Solutions RCP632000BRN
Encompass_Preferred
Toilet bowl brush is designed for easy toilet bowl cleaning. Synthetic polypropylene bristles (1-1/2" long) resist stains, abrasion, discoloration and odors. Durable polypropylene is also resistant to solvents, acids, oils, fungus and bacteria growth. 17" long handle is made of durable plastic.
Catalog No. 50-008-29299