Learn More
Selleck Chemical LLC Rubiadin 1-methyl ether-1MG
SDP
Supplier:  Selleck Chemical LLC E06571MG
Encompass_Preferred
Rubiadin-1-methyl ether a natural anthraquinone compound isolated from the root of Morinda officinalis How can inhibit osteoclastic bone resorption through blocking NF- B pathway and may be a promising agent for the prevention and treatment of bone diseases characterized by excessive bone resorption
Catalog No. 50-313-6859