Learn More
Alkali Scientific Rubidium Chloride, 25 G
SDP
Supplier:  Alkali Scientific RUB002PL
Encompass_Preferred
Rubidium Chloride is a high purity inorganic salt commonly used in biochemical, molecular biology, and analytical laboratory applications. It is widely utilized in cell biology and microbiology research, particularly in protocols related to bacterial transformation and membrane permeability studies. In molecular cloning procedures, rubidium chloride can be used in the preparation of competent bacterial cells, enabling efficient uptake of plasmid DNA. The compound is also employed in ion transport research, spectroscopy studies, and various chemical analyses due to the unique properties of rubidium ions. Laboratory grade rubidium chloride offers consistent purity and stability, making it suitable for controlled experimental environments. It is intended for research and development applications in academic, biotechnology, and pharmaceutical laboratories.
Catalog No. 50-006-63205