Learn More
Alkali Scientific Rubidium Chloride, 500 Grams
SDP
Supplier:  Alkali Scientific RUB005CP
Encompass
Rubidium Chloride is an inorganic salt widely used in laboratory applications, particularly in analytical chemistry and biological research. This 500-gram quantity is suitable for various applications requiring rubidium ions, such as in spectroscopy and ion-selective electrodes. Rubidium chloride is used to prepare solutions for studying ion transport, as well as in the preparation of rubidium salts used in research related to alkali metals. As an analytical grade chemical, it ensures high purity and reliability in experiments, offering consistent results in biological assays and chemical analyses. Rubidium chloride is also used in laboratory synthesis of other rubidium compounds for specialized research.
Catalog No. 50-006-61839