Learn More
Strem, An Ascensus Company CAS# 13446-74-7. 5g. Rubidium fluoride, anhydrous (99.8%-Rb). MFCD00011191
SDP
Supplier:  Strem, An Ascensus Company 9337305G
Encompass
CAS# 13446-74-7. 5g. Rubidium fluoride, anhydrous (99.8%-Rb). MFCD00011191. Molecular Weight: 104.47. Molecular Formula: RbF. Color/form: white pwdr. Strem# 93-3730. http://www.strem.com/catalog/v/93-3730/
Catalog No. 50-901-15158