Learn More
Strem, An Ascensus Company CAS# 12026-05-0. 25g. Rubidium hydroxide hydrate, fused solid (99.8%-Rb). MFCD00149835
SDP
Supplier:  Strem, An Ascensus Company 93372125G
Encompass
CAS# 12026-05-0. 25g. Rubidium hydroxide hydrate, fused solid (99.8%-Rb). MFCD00149835. Molecular Weight: 102.48. Molecular Formula: RbOH∙XH2O. Color/form: white pwdr. Strem# 93-3721. http://www.strem.com/catalog/v/93-3721/
Catalog No. 50-901-15153