Learn More
Medchemexpress LLC Ruboxistaurin | 169939-94-0 | 99.5% | 468.55 g/mol | C28H28N4O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY101955MG
Encompass_Preferred
Ruboxistaurin is a selective, orally active inhibitor of protein kinase C beta (PKCβ) commonly used in research to study signaling pathways and vascular complications. It is supplied as a high-purity, research-grade solid suitable for in vitro and preclinical investigations.
  • Selective inhibitor of protein kinase C beta (PKCβ).
  • Orally active small molecule for preclinical and in vitro research.
  • High reported purity suitable for analytical and biological assays.
  • Available in multiple salt forms and small research sizes.
Catalog No. 50-002-45620