Learn More
Medchemexpress LLC Ruboxistaurin | 169939-94-0 | MFCD09970504 | 99.5% | 468.6 g/mol | C28H28N4O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1019550MG
Encompass_Preferred
Ruboxistaurin is an orally active, selective protein kinase C beta (PKCβ) inhibitor used in research to study vascular complications of diabetes and PKCβ-mediated signaling. It is supplied as a characterized solid for laboratory and preclinical studies.
  • Selective PKCβ inhibition for targeted pathway studies.
  • Orally active pharmacology suitable for in vivo models.
  • High purity (approximately 99.5%) for reproducible results.
  • Well-characterized identifiers: CAS 169939-94-0; formula C28H28N4O3; MW 468.6 g/mol.
  • Brown to reddish brown solid, convenient for formulation and storage.
Catalog No. 50-002-45350