Learn More
Selleck Chemical LLC Ruboxistaurin
SDP
Supplier:  Selleck Chemical LLC S766325MG
Encompass_Preferred
Ruboxistaurin (LY333531) HCl is a -specific protein kinase C inhibitor It competitively and reversibly inhibits PKC 1 and PKC 2 with IC50 values of 4 7 and 5 9 nM respectively
Catalog No. 50-000-03584