Learn More
Medchemexpress LLC Ruboxistaurin hydrochloride | 169939-93-9 | 99.9% | 505.01 g/mol | C28H29ClN4O3 | 50 MG

Supplier: Medchemexpress LLC HY10195B50MG
Ruboxistaurin hydrochloride is the hydrochloride salt of ruboxistaurin, an orally active, selective protein kinase C beta (PKCβ) inhibitor supplied as a solid research reagent for biochemical and cell-based studies. It is intended for research use only and is not for human or clinical applications.
- Selective PKCβ inhibitor with low nanomolar potency (Ki ≈ 2 nM).
- High reported purity (≈99.9%).
- Orange to red solid appearance.
- Molecular weight 505.01 g/mol.
- Stable when stored sealed at 4°C; in solvent -80°C for up to 6 months.
- Suitable for biochemical and cell-based assays probing PKCβ signaling.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.