Learn More
Medchemexpress LLC Ruboxistaurin (LY-333531) | 169939-94-0 | 99.5% | 468.5 g/mol | C28H28N4O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1019510MG
Encompass_Preferred
Ruboxistaurin (LY-333531) is a small-molecule, orally active, selective protein kinase C beta (PKCβ) inhibitor supplied for research use. It is intended for use in biochemical and cell-based studies to probe PKCβ-dependent signaling and exhibits low-nanomolar potency (Ki ≈ 2 nM; IC50 PKCβ1 ≈ 4.7 nM, PKCβ2 ≈ 5.9 nM). Provided as a measured solid for laboratory research; not for human or clinical use.
  • Selective inhibition of protein kinase C beta with low-nanomolar potency.
  • High purity as supplied (HPLC: 99.48%).
  • Suitable for biochemical and cell-based assays.
  • Available in small measured quantities for research use.
  • Soluble in common organic solvents for assay preparation.
Catalog No. 50-002-48619