Learn More
Medchemexpress LLC Ruboxistaurin mesylate | 192050-59-2 | MFCD09970504 | 99.9% | 564.65 g·mol-1 | C29H32N4O6S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10195A100MG
Encompass_Preferred
Ruboxistaurin mesylate is the mesylate salt of ruboxistaurin (LY333531), a selective, orally active ATP-competitive inhibitor of protein kinase C beta (PKCβ) used in preclinical research to probe PKCβ-mediated signaling. The compound is supplied as a stable salt with high reported purity and is commonly prepared in DMSO for assay use.
  • Selective ATP-competitive PKCβ inhibitor with IC50 4.7-5.9 nM.
  • Mesylate salt form for improved stability and handling.
  • High purity suitable for research applications (~99.9%).
  • Soluble in DMSO for in vitro pharmacology studies.
  • Available in laboratory pack sizes such as 100 mg for research use.
Catalog No. 50-002-45867