Learn More
AdipoGen Rubratoxin A (500 µg)
SDP
Supplier:  AdipoGen AGCN20092C500
Encompass_Preferred
Chemical. CAS: 22467-31-8. Formula: C26H32O11. MW: 520.2. Isolated from Penicillium sp. Mycotoxin. Potent and selective competitive protein phosphatase 2A (PP2A) inhibitor. Does not inhibit PP1, PP2B, PTP1B, ALP and CIP. Antitumor agent. Antimetastatic.
Catalog No. 50-205-3452