Learn More
AdipoGen Rubrofusarin
SDP
Supplier:  AdipoGen BVT0395M005
Encompass_Preferred
Chemical. CAS 3567-00-8. Formula C15H12O5. MW 272.3. Isolated from Fusarium graminearum. Monomer of aurofusarin. Mycotoxin. Anticancer and antimycobacterial compound in vivo. Moderate tyrosinase inhibitor. Anti-estrogenic.
Catalog No. 50-256-1950